Learn More
Biolegend APC anti-human CD66b
Supplier:  Biolegend 305117
Encompass_Preferred
APC anti-human CD66b [G10F5]; Isotype: Mouse IgM, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC; Size: 25 tests
Catalog No. NC1562846