Learn More
Beckman Coulter APC Alexa Fluor 750, A78838
Supplier:  Beckman Coulter A78838
Encompass
The Alexa Fluor 750 APC Antibody Labeling Kit from Beckman Coulter is used for labeling antibodies with Alexa Fluor 750 dye for flow cytometry applications. The kit contains reagents for labeling 100-500 μg of antibody. Alexa Fluor 750 dye has an excitation maximum of 749 nm and emission maximum of 775 nm, making it compatible with common flow cytometry lasers and filters. The APC-conjugated antibody is easy to use and stable, providing bright fluorescence intensity. This labeling kit enables multicolor flow cytometry analysis using Alexa Fluor 750 dye.
Catalog No. NC1222697