Learn More
Biolegend APC anti-complement C3b/iC3b
Supplier:  Biolegend 846105
Encompass_Preferred
APC anti-complement C3b/iC3b [3E7/C3b]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Non-Human Primate; Apps: FC; Size: 25 tests
Catalog No. 50-207-4492