Learn More
Biolegend APC anti-DYKDDDDK Tag
Supplier:  Biolegend 637308
Encompass_Preferred
APC anti-DYKDDDDK Tag [L5]; Isotype: Rat IgG2a, λ; Reactivity: DYKDDDDK tag epitope; Apps: FC, ICFC; Size: 100 ug
Catalog No. NC0459005