Learn More
Biolegend APC anti-HA.11 Epitope Tag
Supplier:  Biolegend 901524
Encompass_Preferred
APC anti-HA.11 Epitope Tag [16B12]; Isotype: Mouse IgG1, κ; Reactivity: YPYDVPDYA Tag; Apps: ICFC; Size: 100 ug
Catalog No. NC1889529