Learn More
Biolegend APC anti-human B7-H4
Supplier:  Biolegend 358108
Encompass_Preferred
APC anti-human B7-H4 [MIH43]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC0563159