Learn More
Biolegend APC anti-human CD112R (PVRIG)
Supplier:  Biolegend 301506
Encompass_Preferred
APC anti-human CD112R (PVRIG) [W16216D]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-2775