Learn More
Biolegend APC anti-human CD114 (G-CSFR)
Supplier:  Biolegend 346108
Encompass_Preferred
APC anti-human CD114 (G-CSFR) [LMM741]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cynomolgus; Apps: FC; Size: 100 tests
Catalog No. 50-168-366