Learn More
Biolegend APC anti-human CD132 (common γ chain)
Supplier:  Biolegend 338607
Encompass_Preferred
APC anti-human CD132 (common γ chain) [TUGh4]; Isotype: Rat IgG2b, κ; Reactivity: Human, African Green, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-167-972