Learn More
Biolegend APC anti-human CD138 (Syndecan-1)
Supplier:  Biolegend 352307
Encompass_Preferred
APC anti-human CD138 (Syndecan-1) [DL-101]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus; Apps: FC; Size: 25 tests
Catalog No. 50-168-710