Learn More
Biolegend APC anti-human CD151 (PETA-3)
Supplier:  Biolegend 350405
Encompass_Preferred
APC anti-human CD151 (PETA-3) [50-6]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-168-599