Learn More
Biolegend APC anti-human CD184 (CXCR4)
Supplier:  Biolegend 306509
Encompass_Preferred
APC anti-human CD184 (CXCR4) [12G5]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green, Baboon, Chimpanzee, Cynomolgus, Rhesus, Sooty Mangabey; Apps: FC; Size: 25 tests
Catalog No. 50-166-355