Learn More
Biolegend APC anti-human CD195 (CCR5)
Supplier:  Biolegend 359122
Encompass_Preferred
APC anti-human CD195 (CCR5) [J418F1]; Isotype: Rat IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-850-782