Learn More
Biolegend APC anti-human CD1b
Supplier:  Biolegend 329110
Encompass_Preferred
APC anti-human CD1b [SN13 (K5-1B8)]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Cynomolgus; Apps: FC; Size: 100 tests
Catalog No. 50-207-5839