Learn More
Biolegend APC anti-human CD253 (Trail)
Supplier:  Biolegend 308209
Encompass_Preferred
APC anti-human CD253 (Trail) [RIK-2]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green; Apps: FC; Size: 25 tests
Catalog No. 50-103-0643