Learn More
Biolegend APC anti-human CD254 (TRANCE, RANKL)
Supplier:  Biolegend 347508
Encompass_Preferred
APC anti-human CD254 (TRANCE, RANKL) [MIH24]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-6834