Learn More
Biolegend APC anti-human CD274 (B7-H1, PD-L1)
Supplier:  Biolegend 329708
Encompass_Preferred
APC anti-human CD274 (B7-H1, PD-L1) [29E.2A3]; Isotype: Mouse IgG2b, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC0069310