Learn More
Biolegend APC anti-human CD275 (B7-H2, ICOSL)
Supplier:  Biolegend 309408
Encompass_Preferred
APC anti-human CD275 (B7-H2, ICOSL) [2D3]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-103-0648