Learn More
Biolegend APC anti-human CD28
Supplier:  Biolegend 983406
Encompass_Preferred
APC anti-human CD28 [CD28.2]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 500 uL
Catalog No. 50-207-5570