Learn More
Biolegend APC anti-human CD284 (TLR4)
Supplier:  Biolegend 312815
Encompass_Preferred
APC anti-human CD284 (TLR4) [HTA125]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Cross-Reactivity: Guinea Pig; Apps: FC; Size: 25 tests
Catalog No. 50-604-103