Learn More
Biolegend APC anti-human CD319 (CRACC)
Supplier:  Biolegend 331809
Encompass_Preferred
APC anti-human CD319 (CRACC) [162.1]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cynomolgus; Apps: FC; Size: 25 tests
Catalog No. 50-207-6052