Learn More
Biolegend APC anti-human CD360 (IL-21R)
Supplier:  Biolegend 376106
Encompass_Preferred
APC anti-human CD360 (IL-21R) [W18100A]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-8852