Learn More
Biolegend APC anti-human CD4
Supplier:  Biolegend 300537
Encompass_Preferred
APC anti-human CD4 [RPA-T4]; Isotype: Mouse IgG1, κ; Reactivity: Human, Chimpanzee; Apps: FC; Size: 500 tests
Catalog No. 50-603-974