Learn More
Biolegend APC anti-human CD41
Supplier:  Biolegend 303709
Encompass_Preferred
APC anti-human CD41 [HIP8]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Capuchin Monkey, Cynomolgus, Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-166-031