Learn More
Biolegend APC anti-human CD45RO
Supplier:  Biolegend 304210
Encompass_Preferred
APC anti-human CD45RO [UCHL1]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Chimpanzee, Cynomolgus, Common Marmoset; Apps: FC; Size: 100 tests
Catalog No. 50-166-098