Learn More
Biolegend APC anti-human CD55
Supplier:  Biolegend 311311
Encompass_Preferred
APC anti-human CD55 [JS11]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus; Apps: FC; Size: 25 tests
Catalog No. 50-166-622