Learn More
Biolegend APC anti-human CD62L
Supplier:  Biolegend 304809
Encompass_Preferred
APC anti-human CD62L [DREG-56]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Cattle (Bovine, Cow); Apps: FC; Size: 25 tests
Catalog No. 50-166-162