Learn More
Biolegend APC anti-human CD73 (Ecto-5'-nucleotidase)
Supplier:  Biolegend 344005
Encompass_Preferred
APC anti-human CD73 (Ecto-5'-nucleotidase) [AD2]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon; Apps: FC; Size: 25 tests
Catalog No. 50-168-228