Learn More
Biolegend APC anti-human CD9
Supplier:  Biolegend 312107
Encompass_Preferred
APC anti-human CD9 [HI9a]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cattle (Bovine, Cow), Cynomolgus, Dog (Canine), Horse (Equine), Rabbit (Lapine), Rhesus, Sheep (Ovine); Apps: FC; Size: 25 tests
Catalog No. 50-207-3542