Learn More
Biolegend APC anti-human GARP (LRRC32)
Supplier:  Biolegend 352506
Encompass_Preferred
APC anti-human GARP (LRRC32) [7B11]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-168-722