Learn More
Biolegend APC anti-human Ig light chain κ
Supplier:  Biolegend 316509
Encompass_Preferred
APC anti-human Ig light chain κ [MHK-49]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cynomolgus; Apps: FC; Size: 25 tests
Catalog No. 50-166-870