Learn More
Biolegend APC anti-human IL-6
Supplier:  Biolegend 501112
Encompass_Preferred
APC anti-human IL-6 [MQ2-13A5]; Isotype: Rat IgG1, κ; Reactivity: Human; Apps: ICFC; Size: 100 tests
Catalog No. 50-169-664