Learn More
Biolegend APC anti-human MCP-1
Supplier:  Biolegend 502612
Encompass_Preferred
APC anti-human MCP-1 [5D3-F7]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus, Rhesus; Apps: ICFC; Size: 100 tests
Catalog No. 50-208-5573