Learn More
Biolegend APC anti-human MICA/MICB
Supplier:  Biolegend 320907
Encompass_Preferred
APC anti-human MICA/MICB [6D4]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-167-051