Learn More
Biolegend APC anti-human/mouse Bcl-6
Supplier:  Biolegend 358505
Encompass_Preferred
APC anti-human/mouse Bcl-6 [7D1]; Isotype: Rat IgG2a, κ; Reactivity: Human, Mouse; Apps: ICFC; Size: 25 tests
Catalog No. 50-207-7757