Learn More
Biolegend APC anti-human/mouse CXCR7
Supplier:  Biolegend 331114
Encompass_Preferred
APC anti-human/mouse CXCR7 [8F11-M16]; Isotype: Mouse IgG2b, κ; Reactivity: human, mouse; Apps: FC, ICFC; Size: 100 tests
Catalog No. 50-167-618