Learn More
Biolegend APC anti-human TCR α/β
Supplier:  Biolegend 306717
Encompass_Preferred
APC anti-human TCR α/β [IP26]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-166-387