Learn More
Biolegend APC anti-human TCR Vα24-Jα18 (iNKT cell)
Supplier:  Biolegend 342907
Encompass_Preferred
APC anti-human TCR Vα24-Jα18 (iNKT cell) [6B11]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-reactivity: Pigtailed Macaque, Sooty Mangabey; Apps: FC; Size: 25 tests
Catalog No. 50-168-152