Learn More
Biolegend APC anti-human CD47
Supplier:  Biolegend 323124
Encompass_Preferred
APC anti-human CD47 [CC2C6]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1437716