Learn More
Biolegend APC anti-human CD86
Supplier:  Biolegend 374207
Encompass_Preferred
APC anti-human CD86 [BU63]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. NC1481477