Learn More
Biolegend APC anti-human CCR10 [6588-5]; Isotype: Armenian Hamster IgG; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Supplier:  Biolegend 341506
Encompass_Preferred
APC anti-human CCR10 [6588-5]; Isotype: Armenian Hamster IgG; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1274597