Learn More
Biolegend APC anti-human CD133
Catalog No. 502722073
Encompass_Preferred
APC anti-human CD133 [clone 7]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-272-2073 Supplier Biolegend Supplier No. 372805
APC anti-human CD133 [clone 7]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests