Learn More
Biolegend APC anti-human CD14
Supplier:  Biolegend 367117
Encompass_Preferred
APC anti-human CD14 [63D3]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. NC1124549