Learn More
Biolegend APC anti-human CD140a (PDGFRα)
Supplier:  Biolegend 323512
Encompass_Preferred
APC anti-human CD140a (PDGFRα) [16A1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1825891