Learn More
Biolegend APC anti-human CD152 (CTLA-4)
Supplier:  Biolegend 369612
Encompass_Preferred
APC anti-human CD152 (CTLA-4) [BNI3]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: ICFC, FC; Size: 100 tests
Catalog No. 50-207-8524