Learn More
Biolegend APC anti-human CD193 (CCR3)
Supplier:  Biolegend 310708
Encompass_Preferred
APC anti-human CD193 (CCR3) [5E8]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-166-552