Learn More
Biolegend APC anti-human CD198 (CCR8)
Supplier:  Biolegend 360610
Encompass_Preferred
APC anti-human CD198 (CCR8) [L263G8]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1787413