Learn More
Biolegend APC anti-human CD224
Supplier:  Biolegend 394306
Encompass_Preferred
APC anti-human CD224 [KF29]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-9101