Learn More
Biolegend APC anti-human CD267 (TACI)
Supplier:  Biolegend 311912
Encompass_Preferred
APC anti-human CD267 (TACI) [1A1]; Isotype: Rat IgG2a, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1319473