Learn More
Biolegend APC anti-human CD298
Supplier:  Biolegend 341706
Encompass_Preferred
APC anti-human CD298 [LNH-94]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1466839